The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305242.1 | NP_001292171.1 | 4 - 5 | 511 | 88 |
NM_144988.3 | NP_659425.1 | 3 - 4 | 474 | 88 | |
NR_131032.1 | 2 - 3 | 322 | 88 | ||
GenBank mRNA | AK289395.1 | - | 3 - 4 | 474 | 88 |
BC011706.2 | - | 3 - 4 | 450 | 88 | |
HY073066.1 | - | 2 - 3 | 286 | 88 |