Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM45A
Entrez Gene ID
55076
Gene Name
transmembrane protein 45A
Gene Aliases
DERP7, DNAPTP4
Chromosome Location
Chr.3: 100492619 - 100577453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018004.2 NP_060474.1 4 - 5 896 136
XM_005247569.2 XP_005247626.1 5 - 6 857 136
XM_017006715.1 XP_016862204.1 5 - 6 1017 136
GenBank mRNA AB013911.1 - 3 - 4 583 136
AK000996.1 - 4 - 5 896 136
AK129789.1 - 4 - 5 746 136
AY450392.1 - 4 - 5 896 136
BC040355.1 - 5 - 6 830 136

View Full Product Details