The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077186.1 | NP_001070654.1 | 20 - 21 | 2534 | 118 |
NM_001145809.1 | NP_001139281.1 | 21 - 22 | 2633 | 118 | |
NM_024729.3 | NP_079005.3 | 19 - 20 | 2510 | 118 | |
XM_006723386.3 | XP_006723449.1 | 20 - 21 | 2595 | 118 | |
XM_011527320.2 | XP_011525622.1 | 20 - 21 | 2612 | 118 | |
XM_011527321.2 | XP_011525623.1 | 19 - 20 | 2588 | 118 | |
XM_011527323.2 | XP_011525625.1 | 20 - 21 | 2602 | 118 | |
GenBank mRNA | AB111886.1 | - | 9 - 10 | 1192 | 118 |
AB290169.1 | - | 20 - 21 | 2603 | 118 | |
AK125082.1 | - | 20 - 21 | 2613 | 118 | |
AK307783.1 | - | 20 - 21 | 2516 | 118 | |
AY165122.1 | - | 19 - 20 | 2510 | 118 | |
BC018933.2 | - | 18 - 19 | 2262 | 118 |