Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC44A4
Entrez Gene ID
80736
Gene Name
solute carrier family 44 member 4
Gene Aliases
C6orf29, CTL4, NG22, TPPT
Chromosome Location
Chr.6: 31863192 - 31879046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178045.1 NP_001171516.1 5 - 6 418 101
NM_025257.2 NP_079533.2 5 - 6 415 101
GenBank mRNA AK027397.1 - 5 - 6 366 101
AK222998.1 - 5 - 6 366 101
AK223013.1 - 5 - 6 366 101
AK300550.1 - 5 - 6 418 101
AK314673.1 - 5 - 6 370 101
BC014659.1 - 5 - 6 354 101
DC300771.1 - 5 - 6 415 101
DC392935.1 - 5 - 6 418 101
KU175229.1 - 5 - 6 349 101

View Full Product Details