Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
Chromosome Location
Chr.2: 85354394 - 85391752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135021.1 NP_001128493.1 10 - 11 1148 67
NM_001135022.1 NP_001128494.1 9 - 10 964 67
NM_001135023.1 NP_001128495.1 8 - 9 920 67
NM_032213.4 NP_115589.2 7 - 8 825 67
XM_005264596.3 XP_005264653.1 9 - 10 964 67
XM_005264597.3 XP_005264654.1 8 - 9 1246 67
XM_005264598.3 XP_005264655.1 8 - 9 869 67
XM_017005078.1 XP_016860567.1 8 - 9 820 67
XM_017005079.1 XP_016860568.1 7 - 8 776 67
GenBank mRNA BC001942.1 - 7 - 8 742 67
BC010991.2 - 9 - 10 896 67
BC018666.2 - 7 - 8 742 67
BC054504.1 - 7 - 8 2692 67
BC112324.1 - 8 - 9 883 67
DQ256727.1 - 6 - 7 491 67

View Full Product Details