The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017982.3 | NP_060452.3 | 6 - 7 | 1096 | 90 |
XM_005273169.1 | XP_005273226.1 | 7 - 8 | 1119 | 90 | |
XM_005273172.1 | XP_005273229.1 | 7 - 8 | 1176 | 90 | |
XM_006711408.1 | XP_006711471.1 | 6 - 7 | 1100 | 90 | |
XM_011509685.1 | XP_011507987.1 | 6 - 7 | 1134 | 90 | |
XM_011509687.1 | XP_011507989.1 | 7 - 8 | 1182 | 90 | |
XM_017001583.1 | XP_016857072.1 | 7 - 8 | 1300 | 90 | |
XM_017001584.1 | XP_016857073.1 | 7 - 8 | 1293 | 90 | |
XM_017001585.1 | XP_016857074.1 | 8 - 9 | 1283 | 90 | |
XM_017001586.1 | XP_016857075.1 | 7 - 8 | 1177 | 90 | |
XM_017001587.1 | XP_016857076.1 | 7 - 8 | 1113 | 90 | |
XM_017001588.1 | XP_016857077.1 | 6 - 7 | 1128 | 90 | |
GenBank mRNA | AK096265.1 | - | 5 - 6 | 1550 | 90 |
AK295553.1 | - | 5 - 6 | 852 | 90 | |
AK296923.1 | - | 7 - 8 | 1176 | 90 | |
AY358495.1 | - | 6 - 7 | 1079 | 90 | |
EF560748.1 | - | 6 - 7 | 1024 | 90 |