The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173500.3 | NP_775771.3 | 9 - 10 | 1255 | 102 |
XM_005254171.4 | XP_005254228.1 | 8 - 9 | 1389 | 102 | |
XM_005254173.4 | XP_005254230.1 | 8 - 9 | 1137 | 102 | |
XM_006720402.3 | XP_006720465.1 | 8 - 9 | 864 | 102 | |
XM_006720403.3 | XP_006720466.1 | 9 - 10 | 1031 | 102 | |
XM_017021950.1 | XP_016877439.1 | 7 - 8 | 880 | 102 | |
GenBank mRNA | AK131372.1 | - | 9 - 10 | 979 | 102 |
BC041876.1 | - | 9 - 10 | 1255 | 102 | |
BC071556.1 | - | 10 - 11 | 1029 | 102 |