The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003150.3 | NP_003141.2 | 8 - 9 | 1016 | 69 |
NM_139276.2 | NP_644805.1 | 8 - 9 | 1038 | 69 | |
NM_213662.1 | NP_998827.1 | 8 - 9 | 930 | 69 | |
XM_005257616.3 | XP_005257673.2 | 8 - 9 | 1029 | 69 | |
XM_005257617.3 | XP_005257674.2 | 8 - 9 | 1024 | 69 | |
XM_011525145.2 | XP_011523447.1 | 8 - 9 | 1029 | 69 | |
XM_011525146.2 | XP_011523448.1 | 8 - 9 | 1029 | 69 | |
XM_017024972.1 | XP_016880461.1 | 8 - 9 | 1024 | 69 | |
XM_017024973.1 | XP_016880462.1 | 8 - 9 | 1029 | 69 | |
XM_017024974.1 | XP_016880463.1 | 8 - 9 | 1024 | 69 | |
XM_017024975.1 | XP_016880464.1 | 8 - 9 | 1017 | 69 | |
XM_017024976.1 | XP_016880465.1 | 8 - 9 | 1017 | 69 | |
GenBank mRNA | AB451232.1 | - | 7 - 8 | 798 | 69 |
AJ012463.1 | - | 7 - 8 | 798 | 69 | |
AK291933.1 | - | 8 - 9 | 1111 | 69 | |
AK297994.1 | - | 7 - 8 | 823 | 69 | |
AK301200.1 | - | 8 - 9 | 946 | 69 | |
AK316139.1 | - | 8 - 9 | 968 | 69 | |
BC000627.2 | - | 8 - 9 | 935 | 69 | |
BC014482.1 | - | 8 - 9 | 930 | 69 | |
L29277.1 | - | 8 - 9 | 1018 | 69 |