Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
Chromosome Location
Chr.2: 99545861 - 100142583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025108.1 NP_001020279.1 13 - 14 1592 76
NM_002285.2 NP_002276.2 13 - 14 1609 76
XM_005263943.3 XP_005264000.2 12 - 13 1486 76
XM_005263945.3 XP_005264002.1 4 - 5 843 76
XM_011511169.2 XP_011509471.1 13 - 14 1589 76
XM_011511170.2 XP_011509472.1 14 - 15 1728 76
XM_011511171.2 XP_011509473.1 13 - 14 1592 76
XM_011511172.2 XP_011509474.1 12 - 13 1544 76
XM_011511173.2 XP_011509475.1 13 - 14 1605 76
XM_011511174.2 XP_011509476.1 13 - 14 1549 76
XM_011511175.2 XP_011509477.1 15 - 16 1839 76
XM_011511176.2 XP_011509478.1 12 - 13 1530 76
XM_011511177.2 XP_011509479.1 13 - 14 1620 76
XM_011511178.2 XP_011509480.1 3 - 4 351 76
XM_011511179.2 XP_011509481.1 3 - 4 348 76
XM_017004085.1 XP_016859574.1 4 - 5 840 76
XM_017004086.1 XP_016859575.1 4 - 5 1321 76
XM_017004087.1 XP_016859576.1 4 - 5 1318 76
GenBank mRNA AK092327.1 - 14 - 15 1682 76
AK290468.1 - 12 - 13 1467 76
AK297407.1 - 14 - 15 1857 76
BC036895.1 - 13 - 14 1554 76
BC136579.1 - 11 - 12 1392 76
BC144266.1 - 12 - 13 1467 76
U34360.1 - 11 - 12 1427 76

View Full Product Details