Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
Chromosome Location
Chr.2: 99545861 - 100142583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025108.1 NP_001020279.1 18 - 19 3196 73
NM_002285.2 NP_002276.2 18 - 19 3213 73
XM_005263943.3 XP_005264000.2 17 - 18 3090 73
XM_005263945.3 XP_005264002.1 9 - 10 2447 73
XM_011511169.2 XP_011509471.1 18 - 19 3193 73
XM_011511170.2 XP_011509472.1 19 - 20 3332 73
XM_011511171.2 XP_011509473.1 18 - 19 3196 73
XM_011511172.2 XP_011509474.1 17 - 18 3148 73
XM_011511173.2 XP_011509475.1 18 - 19 3209 73
XM_011511174.2 XP_011509476.1 18 - 19 3153 73
XM_011511175.2 XP_011509477.1 20 - 21 3443 73
XM_011511176.2 XP_011509478.1 17 - 18 3134 73
XM_011511177.2 XP_011509479.1 18 - 19 3224 73
XM_011511178.2 XP_011509480.1 8 - 9 1955 73
XM_011511179.2 XP_011509481.1 8 - 9 1952 73
XM_017004085.1 XP_016859574.1 9 - 10 2444 73
XM_017004086.1 XP_016859575.1 9 - 10 2925 73
XM_017004087.1 XP_016859576.1 9 - 10 2922 73
GenBank mRNA AB209634.1 - 5 - 6 915 73
BC136579.1 - 16 - 17 2996 73
BC144266.1 - 17 - 18 3071 73
U34360.1 - 16 - 17 3031 73

View Full Product Details