The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002020.4 | NP_002011.2 | 16 - 17 | 2483 | 66 |
NM_182925.4 | NP_891555.2 | 16 - 17 | 2483 | 66 | |
XM_011534478.2 | XP_011532780.1 | 16 - 17 | 2861 | 66 | |
XM_011534484.2 | XP_011532786.1 | 14 - 15 | 2041 | 66 | |
XM_017009263.1 | XP_016864752.1 | 16 - 17 | 2867 | 66 | |
XM_017009264.1 | XP_016864753.1 | 16 - 17 | 2867 | 66 | |
XM_017009265.1 | XP_016864754.1 | 16 - 17 | 2868 | 66 | |
XM_017009266.1 | XP_016864755.1 | 16 - 17 | 2862 | 66 | |
XM_017009267.1 | XP_016864756.1 | 16 - 17 | 2862 | 66 | |
XM_017009268.1 | XP_016864757.1 | 16 - 17 | 2426 | 66 | |
GenBank mRNA | AB209637.1 | - | 10 - 11 | 1499 | 66 |
AK291679.1 | - | 16 - 17 | 2459 | 66 | |
AK309910.1 | - | 16 - 17 | 2459 | 66 | |
AY233382.1 | - | 16 - 17 | 2424 | 66 | |
AY233383.1 | - | 16 - 17 | 2424 | 66 | |
U43143.1 | - | 16 - 17 | 2434 | 66 | |
X68203.1 | - | 16 - 17 | 2423 | 66 | |
X69878.1 | - | 16 - 17 | 2425 | 66 |