Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
Chromosome Location
Chr.5: 180601506 - 180650298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002020.4 NP_002011.2 28 - 29 3883 58
NM_182925.4 NP_891555.2 28 - 29 3883 58
XM_011534478.2 XP_011532780.1 28 - 29 4261 58
XM_011534484.2 XP_011532786.1 26 - 27 3441 58
XM_017009266.1 XP_016864755.1 28 - 29 4262 58
XM_017009267.1 XP_016864756.1 28 - 29 4262 58
XM_017009268.1 XP_016864757.1 28 - 29 3826 58
GenBank mRNA AK291679.1 - 28 - 29 3859 58
AK309910.1 - 28 - 29 3859 58
AY233382.1 - 28 - 29 3824 58
AY233383.1 - 28 - 29 3824 58
U43143.1 - 28 - 29 3834 58
X68203.1 - 28 - 29 3823 58
X69878.1 - 28 - 29 3825 58

View Full Product Details