Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM32
Entrez Gene ID
22954
Gene Name
tripartite motif containing 32
Gene Aliases
BBS11, HT2A, LGMD2H, TATIP
Chromosome Location
Chr.9: 116687302 - 116701300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011518398.2 XP_011516700.1 1 - 2 114 69

View Full Product Details