The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016430.3 | NP_057514.2 | 1 - 2 | 158 | 73 |
NM_153682.2 | NP_710149.1 | 1 - 2 | 158 | 73 | |
GenBank mRNA | AB035742.1 | - | 1 - 2 | 90 | 73 |
AB037162.1 | - | 1 - 2 | 94 | 73 | |
AF216305.1 | - | 1 - 2 | 74 | 73 | |
BC005180.1 | - | 1 - 2 | 39 | 73 | |
BC011007.1 | - | 1 - 2 | 43 | 73 | |
BE275469.1 | - | 1 - 2 | 158 | 73 |