The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014044.5 | NP_054763.2 | 4 - 5 | 697 | 109 |
XM_005263913.4 | XP_005263970.1 | 4 - 5 | 793 | 109 | |
XM_006712403.3 | XP_006712466.1 | 4 - 5 | 793 | 109 | |
XM_006712404.2 | XP_006712467.1 | 4 - 5 | 707 | 109 | |
GenBank mRNA | AF077038.1 | - | 4 - 5 | 649 | 109 |
AK222599.1 | - | 4 - 5 | 663 | 109 | |
AY251534.1 | - | 3 - 4 | 558 | 109 | |
BC017019.1 | - | 4 - 5 | 830 | 109 | |
BC025992.2 | - | 3 - 4 | 1725 | 109 | |
BC039078.1 | - | 3 - 4 | 1701 | 109 | |
BC091481.1 | - | 4 - 5 | 687 | 109 | |
KU178586.1 | - | 3 - 4 | 545 | 109 |