Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRID1
Entrez Gene ID
2894
Gene Name
glutamate ionotropic receptor delta type subunit 1
Gene Aliases
GluD1
Chromosome Location
Chr.10: 85599555 - 86366493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017551.2 NP_060021.1 3 - 4 621 113
XM_011539720.2 XP_011538022.1 3 - 4 618 113
GenBank mRNA AK127168.1 - 3 - 4 606 113
AK293104.1 - 3 - 4 579 113
BC039263.1 - 3 - 4 621 113

View Full Product Details