The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281504.1 | NP_001268433.1 | 4 - 5 | 1041 | 88 |
NM_032885.5 | NP_116274.3 | 4 - 5 | 967 | 88 | |
GenBank mRNA | AJ312332.1 | - | 4 - 5 | 783 | 88 |
AK056210.1 | - | 4 - 5 | 942 | 88 | |
AK223383.1 | - | 4 - 5 | 892 | 88 | |
AK294676.1 | - | 4 - 5 | 1041 | 88 | |
BC068992.1 | - | 4 - 5 | 943 | 88 |