Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG4D
Entrez Gene ID
84971
Gene Name
autophagy related 4D cysteine peptidase
Gene Aliases
APG4-D, APG4D, AUTL4
Chromosome Location
Chr.19: 10543894 - 10553423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281504.1 NP_001268433.1 9 - 10 1517 96
NM_032885.5 NP_116274.3 9 - 10 1443 96
NR_104024.1 9 - 10 1399 96
NR_104025.1 8 - 9 1166 96
XM_006722927.1 XP_006722990.1 7 - 8 1098 96
GenBank mRNA AJ312332.1 - 9 - 10 1259 96
AK223383.1 - 9 - 10 1368 96
AK294676.1 - 9 - 10 1517 96
AK314712.1 - 9 - 10 1378 96
BC007639.1 - 8 - 9 1089 96
BC016845.1 - 4 - 5 401 96
BC068992.1 - 9 - 10 1419 96

View Full Product Details