The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003167.3 | NP_003158.2 | 1 - 2 | 272 | 132 |
GenBank mRNA | AK289380.1 | - | 1 - 2 | 214 | 132 |
AK313415.1 | - | 1 - 2 | 199 | 132 | |
BC020755.1 | - | 1 - 2 | 189 | 132 | |
BG563083.1 | - | 1 - 2 | 252 | 132 | |
BX509490.1 | - | 1 - 2 | 273 | 132 | |
L02337.1 | - | 1 - 2 | 197 | 132 | |
L20000.1 | - | 1 - 2 | 184 | 132 | |
S43859.1 | - | 1 - 2 | 197 | 132 | |
U08024.1 | - | 1 - 2 | 152 | 132 | |
U08025.1 | - | 1 - 2 | 196 | 132 | |
X70222.1 | - | 1 - 2 | 184 | 132 | |
X84816.1 | - | 1 - 2 | 177 | 132 |