Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BACE2
Entrez Gene ID
25825
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location
Chr.21: 41167801 - 41282534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012105.4 NP_036237.2 1 - 2 776 69
NM_138991.2 NP_620476.1 1 - 2 776 69
NM_138992.2 NP_620477.1 1 - 2 776 69
GenBank mRNA AF050171.1 - 1 - 2 421 69
AF117892.1 - 1 - 2 413 69
AF178532.1 - 1 - 2 776 69
AF188276.1 - 1 - 2 776 69
AF188277.1 - 1 - 2 776 69
AF200192.1 - 1 - 2 390 69
AF200342.1 - 1 - 2 313 69
AF204944.1 - 1 - 2 403 69
AF212252.1 - 1 - 2 134 69
AK075539.1 - 2 - 3 270 69
AK292056.1 - 1 - 2 449 69
AY358927.1 - 1 - 2 406 69
BC014453.1 - 1 - 2 404 69

View Full Product Details