Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 14 - 15 2077 105
XM_005252965.4 XP_005253022.1 15 - 16 1769 105
XM_011520157.2 XP_011518459.1 14 - 15 1630 105
XM_011520158.2 XP_011518460.1 15 - 16 1982 105
XM_011520160.2 XP_011518462.1 17 - 18 2143 105
XM_011520161.2 XP_011518463.1 17 - 18 2234 105
XM_011520162.2 XP_011518464.1 17 - 18 2112 105
XM_011520164.2 XP_011518466.1 16 - 17 2041 105
XM_011520165.2 XP_011518467.1 16 - 17 1996 105
XM_011520166.2 XP_011518468.1 16 - 17 1919 105
XM_011520167.2 XP_011518469.1 16 - 17 2122 105
XM_011520168.2 XP_011518470.1 16 - 17 1974 105
XM_011520169.2 XP_011518471.1 15 - 16 1933 105
XM_011520173.2 XP_011518475.1 15 - 16 1889 105
XM_011520174.2 XP_011518476.1 16 - 17 1863 105
XM_011520175.2 XP_011518477.1 16 - 17 1860 105
XM_017017885.1 XP_016873374.1 17 - 18 2062 105
XM_017017886.1 XP_016873375.1 16 - 17 2027 105
XM_017017887.1 XP_016873376.1 15 - 16 1807 105
XM_017017888.1 XP_016873377.1 15 - 16 1979 105
XM_017017889.1 XP_016873378.1 15 - 16 1979 105
XM_017017890.1 XP_016873379.1 15 - 16 1976 105
XM_017017891.1 XP_016873380.1 15 - 16 1886 105
XM_017017892.1 XP_016873381.1 15 - 16 1886 105
XM_017017893.1 XP_016873382.1 15 - 16 1982 105
XM_017017897.1 XP_016873386.1 16 - 17 2059 105
XM_017017898.1 XP_016873387.1 16 - 17 1917 105
XM_017017899.1 XP_016873388.1 17 - 18 2051 105
XM_017017900.1 XP_016873389.1 16 - 17 2006 105
XM_017017904.1 XP_016873393.1 15 - 16 1982 105
XM_017017905.1 XP_016873394.1 14 - 15 1730 105
XM_017017907.1 XP_016873396.1 14 - 15 1637 105
XM_017017908.1 XP_016873397.1 17 - 18 2150 105
GenBank mRNA BC015714.2 - 15 - 16 1741 105
BX648236.1 - 7 - 8 2311 105

View Full Product Details