The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145415.1 | NP_001138887.1 | 2 - 3 | 447 | 78 |
NM_001243491.1 | NP_001230420.1 | 2 - 3 | 393 | 78 | |
NM_012432.3 | NP_036564.3 | 2 - 3 | 447 | 78 | |
XM_005245641.3 | XP_005245698.1 | 2 - 3 | 447 | 78 | |
XM_017002953.1 | XP_016858442.1 | 2 - 3 | 371 | 78 | |
XM_017002954.1 | XP_016858443.1 | 2 - 3 | 447 | 78 | |
XM_017002955.1 | XP_016858444.1 | 2 - 3 | 455 | 78 | |
GenBank mRNA | AK308335.1 | - | 2 - 3 | 391 | 78 |
BC009362.2 | - | 1 - 2 | 283 | 78 | |
BC028671.2 | - | 2 - 3 | 374 | 78 | |
D31891.1 | - | 2 - 3 | 343 | 78 | |
DA418593.1 | - | 2 - 3 | 447 | 78 | |
DB062762.1 | - | 2 - 3 | 383 | 78 |