The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171162.1 | NP_001164633.1 | 24 - 25 | 4638 | 87 |
NM_005096.3 | NP_005087.1 | 24 - 25 | 4113 | 87 | |
NM_201599.2 | NP_963893.1 | 24 - 25 | 4104 | 87 | |
XM_005262309.3 | XP_005262366.1 | 24 - 25 | 4547 | 87 | |
XM_005262310.2 | XP_005262367.1 | 24 - 25 | 4024 | 87 | |
XM_011531062.2 | XP_011529364.1 | 24 - 25 | 4013 | 87 | |
GenBank mRNA | AB002383.1 | - | 24 - 25 | 4012 | 87 |
BC069057.1 | - | 24 - 25 | 4017 | 87 | |
X95808.1 | - | 25 - 26 | 4614 | 87 |