The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171162.1 | NP_001164633.1 | 14 - 15 | 3121 | 64 |
XM_005262310.2 | XP_005262367.1 | 14 - 15 | 2507 | 64 | |
GenBank mRNA | X95808.1 | - | 15 - 16 | 3097 | 64 |