The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002504.5 | NP_002495.2 | 11 - 12 | 2187 | 72 |
NM_147134.3 | NP_667345.1 | 11 - 12 | 2187 | 72 | |
GenBank mRNA | AF332009.1 | - | 11 - 12 | 2092 | 72 |
AK225750.1 | - | 11 - 12 | 2060 | 72 | |
BC006447.1 | - | 10 - 11 | 1684 | 72 | |
BC012151.1 | - | 11 - 12 | 2057 | 72 | |
U15306.1 | - | 10 - 11 | 1983 | 72 |