The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318758.1 | NP_001305687.1 | 13 - 14 | 2378 | 61 |
NM_002504.5 | NP_002495.2 | 13 - 14 | 2375 | 61 | |
NM_147134.3 | NP_667345.1 | 13 - 14 | 2375 | 61 | |
GenBank mRNA | AF332009.1 | - | 13 - 14 | 2280 | 61 |
AK225750.1 | - | 13 - 14 | 2248 | 61 | |
AK291643.1 | - | 13 - 14 | 2268 | 61 | |
AL832342.1 | - | 13 - 14 | 2261 | 61 | |
BC006447.1 | - | 12 - 13 | 1872 | 61 | |
BC012151.1 | - | 13 - 14 | 2245 | 61 | |
U15306.1 | - | 12 - 13 | 2171 | 61 |