The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002035.2 | NP_002026.1 | 8 - 9 | 937 | 70 |
XM_017025690.1 | XP_016881179.1 | 8 - 9 | 1142 | 70 | |
GenBank mRNA | AK025120.1 | - | 8 - 9 | 870 | 70 |
AK126679.1 | - | 2 - 3 | 2210 | 70 | |
AK297670.1 | - | 7 - 8 | 676 | 70 | |
AK312360.1 | - | 8 - 9 | 865 | 70 | |
BC008797.2 | - | 8 - 9 | 916 | 70 | |
BT006782.1 | - | 8 - 9 | 782 | 70 | |
X63657.1 | - | 8 - 9 | 889 | 70 |