The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007189.1 | NP_001007190.1 | 1 - 1 | 2642 | 73 |
GenBank mRNA | AB073888.1 | - | 1 - 1 | 958 | 73 |
BC017422.1 | - | 1 - 1 | 234 | 73 | |
BC041380.1 | - | 1 - 1 | 2646 | 73 |