The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172630.1 | NP_001166101.1 | 18 - 19 | 1988 | 103 |
XM_005258488.1 | XP_005258545.1 | 19 - 20 | 2082 | 103 | |
XM_006722999.1 | XP_006723062.1 | 19 - 20 | 2115 | 103 | |
XM_006723003.2 | XP_006723066.1 | 4 - 5 | 615 | 103 | |
XM_011526416.1 | XP_011524718.1 | 18 - 19 | 1970 | 103 | |
XM_011526417.1 | XP_011524719.1 | 18 - 19 | 1943 | 103 | |
XM_011526418.1 | XP_011524720.1 | 18 - 19 | 1973 | 103 | |
XM_011526419.1 | XP_011524721.1 | 18 - 19 | 1940 | 103 | |
XM_011526420.1 | XP_011524722.1 | 18 - 19 | 2021 | 103 | |
GenBank mRNA | AK127255.1 | - | 18 - 19 | 1988 | 103 |
AL137579.1 | - | 6 - 7 | 668 | 103 | |
BC128586.1 | - | 19 - 20 | 1999 | 103 |