Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
Chromosome Location
Chr.11: 47659168 - 47715397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005253141.4 XP_005253198.1 8 - 9 2270 110
GenBank mRNA BC028200.1 - 1 - 2 301 110

View Full Product Details