Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBL3
Entrez Gene ID
10607
Gene Name
transducin beta like 3
Gene Aliases
SAZD, UTP13
Chromosome Location
Chr.16: 1972063 - 1978750 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006453.2 NP_006444.2 15 - 16 1770 95
GenBank mRNA AB209173.1 - 15 - 16 1742 95
AK095076.1 - 7 - 8 1946 95
BC007620.1 - 7 - 8 954 95
BC010231.1 - 15 - 16 1770 95
BC014007.2 - 15 - 16 1742 95
BC035409.1 - 15 - 16 1726 95

View Full Product Details