Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBL3
Entrez Gene ID
10607
Gene Name
transducin beta like 3
Gene Aliases
SAZD, UTP13
Chromosome Location
Chr.16: 1972063 - 1978750 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006453.2 NP_006444.2 5 - 6 485 76
GenBank mRNA AB209173.1 - 5 - 6 457 76
BC010231.1 - 5 - 6 485 76
BC014007.2 - 5 - 6 457 76
BC035409.1 - 5 - 6 441 76
U02609.1 - 5 - 6 433 76

View Full Product Details