Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBL3
Entrez Gene ID
10607
Gene Name
transducin beta like 3
Gene Aliases
SAZD, UTP13
Chromosome Location
Chr.16: 1972063 - 1978750 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006453.2 NP_006444.2 9 - 10 913 112
GenBank mRNA AB209173.1 - 9 - 10 885 112
AK095076.1 - 4 - 5 388 112
BC007620.1 - 1 - 2 97 112
BC010231.1 - 9 - 10 913 112
BC014007.2 - 9 - 10 885 112
BC035409.1 - 9 - 10 869 112
U02609.1 - 9 - 10 861 112

View Full Product Details