The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006453.2 | NP_006444.2 | 2 - 3 | 194 | 64 |
GenBank mRNA | AB209173.1 | - | 2 - 3 | 166 | 64 |
BC010231.1 | - | 2 - 3 | 194 | 64 | |
BC014007.2 | - | 2 - 3 | 166 | 64 | |
BC035409.1 | - | 2 - 3 | 150 | 64 | |
U02609.1 | - | 2 - 3 | 144 | 64 |