The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135155.2 | NP_001128627.1 | 9 - 10 | 1002 | 134 |
NM_001135156.2 | NP_001128628.1 | 9 - 10 | 1016 | 134 | |
NM_001289978.1 | NP_001276907.1 | 9 - 10 | 1002 | 134 | |
NM_004647.3 | NP_004638.2 | 8 - 9 | 870 | 134 | |
XM_005259289.2 | XP_005259346.1 | 8 - 9 | 951 | 134 | |
XM_005259290.3 | XP_005259347.1 | 9 - 10 | 973 | 134 | |
XM_005259292.4 | XP_005259349.1 | 8 - 9 | 888 | 134 | |
XM_006723407.2 | XP_006723470.1 | 9 - 10 | 973 | 134 | |
XM_006723408.3 | XP_006723471.1 | 9 - 10 | 941 | 134 | |
XM_006723409.2 | XP_006723472.1 | 9 - 10 | 988 | 134 | |
XM_006723410.2 | XP_006723473.1 | 6 - 7 | 540 | 134 | |
XM_011527356.1 | XP_011525658.1 | 8 - 9 | 951 | 134 | |
XM_011527357.2 | XP_011525659.1 | 8 - 9 | 891 | 134 | |
XM_011527358.2 | XP_011525660.1 | 9 - 10 | 1017 | 134 | |
GenBank mRNA | AK094632.1 | - | 9 - 10 | 1329 | 134 |
AK127198.1 | - | 5 - 6 | 1682 | 134 | |
BC021191.2 | - | 9 - 10 | 975 | 134 | |
BC125152.1 | - | 8 - 9 | 1110 | 134 | |
BC125153.1 | - | 9 - 10 | 904 | 134 | |
GQ358728.1 | - | 9 - 10 | 894 | 134 | |
U43843.1 | - | 8 - 9 | 768 | 134 |