The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287489.1 | NP_001274418.1 | 23 - 24 | 2994 | 67 |
NM_004802.3 | NP_004793.2 | 6 - 7 | 852 | 67 | |
NM_194248.2 | NP_919224.1 | 23 - 24 | 2994 | 67 | |
NM_194322.2 | NP_919303.1 | 5 - 6 | 946 | 67 | |
NM_194323.2 | NP_919304.1 | 6 - 7 | 852 | 67 | |
XM_017005338.1 | XP_016860827.1 | 23 - 24 | 2867 | 67 | |
GenBank mRNA | AF107403.1 | - | 6 - 7 | 852 | 67 |
AF183185.1 | - | 23 - 24 | 2994 | 67 | |
AF183186.1 | - | 5 - 6 | 946 | 67 | |
AF183187.1 | - | 6 - 7 | 852 | 67 |