Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOF
Entrez Gene ID
9381
Gene Name
otoferlin
Gene Aliases
AUNB1, DFNB6, DFNB9, FER1L2, NSRD9
Chromosome Location
Chr.2: 26457203 - 26558698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287489.1 NP_001274418.1 33 - 34 4215 68
NM_004802.3 NP_004793.2 16 - 17 2013 68
NM_194248.2 NP_919224.1 33 - 34 4215 68
NM_194322.2 NP_919303.1 15 - 16 2167 68
NM_194323.2 NP_919304.1 16 - 17 2013 68
XM_017005338.1 XP_016860827.1 33 - 34 4028 68
GenBank mRNA AF107403.1 - 16 - 17 2013 68
AF183185.1 - 33 - 34 4215 68
AF183186.1 - 15 - 16 2167 68
AF183187.1 - 16 - 17 2013 68
AK296272.1 - 6 - 7 570 68

View Full Product Details