The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287489.1 | NP_001274418.1 | 36 - 37 | 4626 | 71 |
NM_004802.3 | NP_004793.2 | 19 - 20 | 2424 | 71 | |
NM_194248.2 | NP_919224.1 | 36 - 37 | 4626 | 71 | |
NM_194322.2 | NP_919303.1 | 18 - 19 | 2578 | 71 | |
NM_194323.2 | NP_919304.1 | 19 - 20 | 2424 | 71 | |
XM_017005338.1 | XP_016860827.1 | 36 - 37 | 4439 | 71 | |
GenBank mRNA | AF107403.1 | - | 19 - 20 | 2424 | 71 |
AF183185.1 | - | 36 - 37 | 4626 | 71 | |
AF183186.1 | - | 18 - 19 | 2578 | 71 | |
AF183187.1 | - | 19 - 20 | 2424 | 71 | |
AK296272.1 | - | 9 - 10 | 981 | 71 |