The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017553.2 | NP_060023.1 | 4 - 5 | 696 | 151 |
NR_104038.1 | 4 - 5 | 696 | 151 | ||
XM_011521685.2 | XP_011519987.1 | 4 - 5 | 1531 | 151 | |
GenBank mRNA | AB033085.1 | - | 4 - 5 | 592 | 151 |
AK290371.1 | - | 3 - 4 | 434 | 151 | |
BC144681.1 | - | 4 - 5 | 582 | 151 | |
BC146785.1 | - | 4 - 5 | 604 | 151 |