Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTWD1
Entrez Gene ID
56986
Gene Name
DTW domain containing 1
Gene Aliases
MDS009
Chromosome Location
Chr.15: 49620980 - 49654448 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017022419.1 XP_016877908.1 1 - 2 140 104
XM_017022428.1 XP_016877917.1 1 - 2 105 104

View Full Product Details