The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040450.2 | NP_001035540.1 | 7 - 8 | 1740 | 112 |
NM_001040453.2 | NP_001035543.1 | 7 - 8 | 1570 | 112 | |
XM_011521687.1 | XP_011519989.1 | 8 - 9 | 1792 | 112 | |
XM_011521688.1 | XP_011519990.1 | 8 - 9 | 1792 | 112 | |
XM_011521689.1 | XP_011519991.1 | 8 - 9 | 1690 | 112 | |
XM_011521690.1 | XP_011519992.1 | 7 - 8 | 1636 | 112 | |
XM_017022332.1 | XP_016877821.1 | 6 - 7 | 1546 | 112 | |
XM_017022333.1 | XP_016877822.1 | 5 - 6 | 1492 | 112 | |
XM_017022334.1 | XP_016877823.1 | 5 - 6 | 1492 | 112 | |
GenBank mRNA | AB032990.1 | - | 7 - 8 | 847 | 112 |
AK075319.1 | - | 7 - 8 | 1618 | 112 | |
AK297291.1 | - | 7 - 8 | 1618 | 112 | |
AL833610.1 | - | 6 - 7 | 1598 | 112 | |
BC140811.1 | - | 7 - 8 | 1570 | 112 | |
BC144616.1 | - | 7 - 8 | 1570 | 112 |