Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAT3
Entrez Gene ID
6774
Gene Name
signal transducer and activator of transcription 3
Gene Aliases
ADMIO, ADMIO1, APRF, HIES
Chromosome Location
Chr.17: 42313324 - 42388505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003150.3 NP_003141.2 24 - 24 4416 153
NM_139276.2 NP_644805.1 24 - 24 4441 153
NM_213662.1 NP_998827.1 24 - 24 4283 153
XM_011525145.2 XP_011523447.1 24 - 24 4432 153
XM_011525146.2 XP_011523448.1 24 - 24 4382 153
XM_017024973.1 XP_016880462.1 24 - 24 4379 153
GenBank mRNA AK024535.1 - 1 - 1 1006 153
BC008044.2 - 1 - 1 913 153
BC067119.1 - 1 - 1 561 153
CF454565.1 - 1 - 1 559 153

View Full Product Details