Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAT3
Entrez Gene ID
6774
Gene Name
signal transducer and activator of transcription 3
Gene Aliases
ADMIO, ADMIO1, APRF, HIES
Chromosome Location
Chr.17: 42313324 - 42388505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003150.3 NP_003141.2 6 - 7 770 97
NM_139276.2 NP_644805.1 6 - 7 792 97
NM_213662.1 NP_998827.1 6 - 7 684 97
XM_005257616.3 XP_005257673.2 6 - 7 783 97
XM_005257617.3 XP_005257674.2 6 - 7 778 97
XM_011525145.2 XP_011523447.1 6 - 7 783 97
XM_011525146.2 XP_011523448.1 6 - 7 783 97
XM_017024972.1 XP_016880461.1 6 - 7 778 97
XM_017024973.1 XP_016880462.1 6 - 7 783 97
XM_017024974.1 XP_016880463.1 6 - 7 778 97
XM_017024975.1 XP_016880464.1 6 - 7 771 97
XM_017024976.1 XP_016880465.1 6 - 7 771 97
GenBank mRNA AB451232.1 - 5 - 6 552 97
AJ012463.1 - 5 - 6 552 97
AK291933.1 - 6 - 7 865 97
AK297994.1 - 5 - 6 577 97
AK301200.1 - 6 - 7 700 97
AK316139.1 - 6 - 7 722 97
BC000627.2 - 6 - 7 689 97
BC014482.1 - 6 - 7 684 97
L29277.1 - 6 - 7 772 97

View Full Product Details