Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K3
Entrez Gene ID
4215
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
MAPKKK3, MEKK3
Chromosome Location
Chr.17: 63622209 - 63696310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002401.3 NP_002392.2 12 - 13 1529 94
NM_203351.1 NP_976226.1 13 - 14 1622 94
XM_005257376.3 XP_005257433.1 13 - 14 1842 94
XM_005257377.3 XP_005257434.1 12 - 13 1241 94
XM_005257378.2 XP_005257435.1 13 - 14 1854 94
GenBank mRNA AK225784.1 - 13 - 14 1626 94
AK308152.1 - 7 - 8 738 94
AK315305.1 - 13 - 14 1308 94
AL834303.1 - 13 - 14 1621 94
AX775739.1 - 14 - 15 1369 94
AX775741.1 - 14 - 15 1369 94
BC008336.1 - 11 - 12 1114 94
BC090859.1 - 12 - 13 1530 94
BC093672.1 - 12 - 13 1260 94
BC093674.1 - 12 - 13 1260 94
BC143224.1 - 12 - 13 1248 94
BC143235.1 - 12 - 13 1257 94
DQ454151.1 - 11 - 12 1198 94
U78876.1 - 12 - 13 1293 94

View Full Product Details