The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002401.3 | NP_002392.2 | 8 - 9 | 1032 | 74 |
NM_203351.1 | NP_976226.1 | 9 - 10 | 1125 | 74 | |
XM_005257378.2 | XP_005257435.1 | 9 - 10 | 1357 | 74 | |
GenBank mRNA | AK315305.1 | - | 9 - 10 | 811 | 74 |
AL834303.1 | - | 9 - 10 | 1124 | 74 | |
AX775739.1 | - | 10 - 11 | 872 | 74 | |
AX775741.1 | - | 10 - 11 | 872 | 74 | |
BC008336.1 | - | 7 - 8 | 617 | 74 | |
BC090859.1 | - | 8 - 9 | 1033 | 74 | |
BC093672.1 | - | 8 - 9 | 763 | 74 | |
BC093674.1 | - | 8 - 9 | 763 | 74 | |
BC143235.1 | - | 8 - 9 | 760 | 74 | |
U78876.1 | - | 8 - 9 | 796 | 74 |