The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001359.1 | NP_001350.1 | 8 - 9 | 1024 | 80 |
XM_005250808.4 | XP_005250865.1 | 7 - 8 | 712 | 80 | |
XM_011516880.2 | XP_011515182.1 | 9 - 10 | 1019 | 80 | |
XM_017013147.1 | XP_016868636.1 | 10 - 11 | 1679 | 80 | |
XM_017013148.1 | XP_016868637.1 | 10 - 11 | 1419 | 80 | |
GenBank mRNA | AK300069.1 | - | 10 - 11 | 1539 | 80 |
BC105080.1 | - | 8 - 9 | 971 | 80 | |
BC105082.1 | - | 8 - 9 | 971 | 80 | |
BC143669.1 | - | 6 - 7 | 668 | 80 | |
L26050.1 | - | 8 - 9 | 893 | 80 |