Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYBU
Entrez Gene ID
55638
Gene Name
syntabulin
Gene Aliases
GOLSYN, OCSYN, SNPHL
Chromosome Location
Chr.8: 109574176 - 109691791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099743.1 NP_001093213.1 4 - 5 946 86
NM_001099744.1 NP_001093214.1 4 - 5 949 86
NM_001099745.1 NP_001093215.1 4 - 5 785 86
NM_001099746.1 NP_001093216.1 2 - 3 499 86
NM_001099747.1 NP_001093217.1 4 - 5 703 86
NM_001099748.1 NP_001093218.1 4 - 5 706 86
NM_001099749.1 NP_001093219.1 2 - 3 420 86
NM_001099750.1 NP_001093220.1 5 - 6 827 86
NM_001099751.1 NP_001093221.1 4 - 5 792 86
NM_001099752.1 NP_001093222.1 4 - 5 795 86
NM_001099753.1 NP_001093223.1 3 - 4 593 86
NM_001099754.1 NP_001093224.1 3 - 4 596 86
NM_001099755.1 NP_001093225.1 2 - 3 391 86
NM_001099756.1 NP_001093226.1 2 - 3 667 86
NM_017786.5 NP_060256.3 5 - 6 824 86
XM_017013614.1 XP_016869103.1 3 - 4 515 86
XM_017013615.1 XP_016869104.1 3 - 4 489 86
GenBank mRNA AB040905.1 - 2 - 3 249 86
AB195676.1 - 5 - 6 757 86
AB195677.1 - 4 - 5 636 86
AB195678.1 - 2 - 3 353 86
AB195679.1 - 2 - 3 1152 86
AK000373.1 - 2 - 3 478 86
AK055246.1 - 4 - 5 716 86
AK090559.1 - 2 - 3 386 86
AK096558.1 - 3 - 4 591 86
AK290469.1 - 4 - 5 785 86
AL713645.1 - 5 - 6 718 86
AY705385.1 - 3 - 4 433 86
BC012082.1 - 2 - 3 366 86
DA530258.1 - 2 - 3 420 86
DB127917.1 - 2 - 3 391 86
DB155885.1 - 2 - 3 460 86

View Full Product Details