The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002745.4 | NP_002736.3 | 8 - 9 | 1334 | 66 |
GenBank mRNA | AK090869.1 | - | 9 - 10 | 1191 | 66 |
AK295188.1 | - | 10 - 11 | 1267 | 66 | |
AK307615.1 | - | 9 - 10 | 1179 | 66 | |
BC099905.1 | - | 8 - 9 | 1299 | 66 | |
Z11694.1 | - | 8 - 9 | 1278 | 66 | |
Z11695.1 | - | 8 - 9 | 1192 | 66 |