The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000192.3 | NP_000183.2 | 7 - 8 | 1422 | 100 |
NM_080717.2 | NP_542448.1 | 6 - 7 | 1237 | 100 | |
NM_181486.2 | NP_852259.1 | 7 - 8 | 1329 | 100 | |
XM_017019912.1 | XP_016875401.1 | 7 - 8 | 3173 | 100 | |
GenBank mRNA | AB051068.1 | - | 6 - 7 | 1182 | 100 |
AF221714.1 | - | 7 - 8 | 1420 | 100 | |
AK290611.1 | - | 7 - 8 | 926 | 100 | |
AK300251.1 | - | 5 - 6 | 723 | 100 | |
BC027942.1 | - | 7 - 8 | 1250 | 100 | |
U80987.1 | - | 6 - 7 | 755 | 100 | |
U89353.1 | - | 6 - 7 | 755 | 100 | |
Y09445.1 | - | 7 - 8 | 1411 | 100 |