Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POPDC2
Entrez Gene ID
64091
Gene Name
popeye domain containing 2
Gene Aliases
POP2
Chromosome Location
Chr.3: 119642052 - 119660590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK377180.1 - 4 - 5 411 110
AK377184.1 - 4 - 5 411 110
BC026911.2 - 1 - 2 69 110

View Full Product Details